Gene Transcripts

Gene Symbol
Fgf16
Entrez Gene ID
80903
Gene Name
fibroblast growth factor 16
Gene Aliases
-
Chromosome Location
Chr.X: 105764477 - 105776532 on Build GRCm38
Uni Gene
Mm.154768
Interrogated Sequence Translated Protein
RefSeq NM_030614.2 NP_085117.2
GenBank mRNA AB049219.1 -
AF292104.1 -
BB342405.2 -
BC132641.1 -
BC137993.1 -

View Full Product Details