Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_008366.3 | NP_032392.1 |
GenBank mRNA | AF065914.1 | - |
AF065915.1 | - | |
AF065916.1 | - | |
AF352786.1 | - | |
AF538059.1 | - | |
AF542383.1 | - | |
AF542384.1 | - | |
AF542385.1 | - | |
AY147902.1 | - | |
BC116845.1 | - | |
BC116873.1 | - | |
BX519775.1 | - | |
DQ836354.1 | - | |
K02292.1 | - | |
U41494.1 | - | |
U41504.1 | - | |
U41505.1 | - | |
U41506.1 | - | |
X01772.1 | - | |
X66058.1 | - | |
X73040.1 | - |