Gene Transcripts

Gene Symbol
Igf1
Entrez Gene ID
16000
Gene Name
insulin-like growth factor 1
Gene Aliases
C730016P09Rik, Igf-1, Igf-I
Chromosome Location
Chr.10: 87858270 - 87937047 on Build GRCm38
Uni Gene
Mm.268521
Interrogated Sequence Translated Protein
RefSeq NM_001111274.1 NP_001104744.1
NM_001111275.1 NP_001104745.1
NM_001111276.1 NP_001104746.1
NM_010512.4 NP_034642.2
NM_184052.3 NP_908941.1
GenBank mRNA AF440694.1 -
AK038119.1 -
AK050118.1 -
AK052033.1 -
AK081019.1 -
AK155435.1 -
AK165471.1 -
AY878192.1 -
AY878193.1 -
BC012409.1 -
CA494190.1 -
CT010364.1 -
X04480.1 -
X04482.1 -

View Full Product Details