Gene Transcripts

Gene Symbol
Adam6a
Entrez Gene ID
238406
Gene Name
a disintegrin and metallopeptidase domain 6A
Gene Aliases
Adam6, tMDCIV
Chromosome Location
Chr.12: 113543908 - 113546414 on Build GRCm38
Uni Gene
Mm.188683
Interrogated Sequence Translated Protein
RefSeq NM_174885.3 NP_777479.2
GenBank mRNA AY158689.1 -
BC048757.1 -
BC108387.1 -
BC139059.1 -
KF144375.1 -
KF144376.1 -

View Full Product Details