Gene Transcripts

Gene Symbol
Il2
Entrez Gene ID
16183
Gene Name
interleukin 2
Gene Aliases
Il-2
Chromosome Location
Chr.3: 37120713 - 37125954 on Build GRCm38
Uni Gene
Mm.14190
Interrogated Sequence Translated Protein
RefSeq NM_008366.3 NP_032392.1
GenBank mRNA AF065914.1 -
AF065915.1 -
AF065916.1 -
AF352786.1 -
AF538059.1 -
AF542383.1 -
AF542384.1 -
AF542385.1 -
AY147902.1 -
BC116845.1 -
BC116873.1 -
BX519775.1 -
DQ836354.1 -
K02292.1 -
U41494.1 -
U41504.1 -
U41505.1 -
U41506.1 -
X01772.1 -
X66058.1 -
X73040.1 -

View Full Product Details