Gene Transcripts

Gene Symbol
Tnf
Entrez Gene ID
21926
Gene Name
tumor necrosis factor
Gene Aliases
DIF, TNF-a, TNF-alpha, TNFSF2, TNFalpha, Tnfa, Tnfsf1a
Chromosome Location
Chr.17: 35199367 - 35202007 on Build GRCm38
Uni Gene
Mm.1293
Interrogated Sequence Translated Protein
RefSeq NM_001278601.1 NP_001265530.1
NM_013693.3 NP_038721.1
GenBank mRNA AK153319.1 -
AK153800.1 -
AK154223.1 -
AK155964.1 -
AY423855.1 -
BC117057.1 -
BC137720.1 -
BY167004.1 -
M11731.1 -
M13049.1 -
X02611.1 -

View Full Product Details