Gene Transcripts

Gene Symbol
Fgf9
Entrez Gene ID
14180
Gene Name
fibroblast growth factor 9
Gene Aliases
Eks
Chromosome Location
Chr.14: 58071037 - 58112720 on Build GRCm38
Uni Gene
Mm.8846
Interrogated Sequence Translated Protein
RefSeq NM_013518.4 NP_038546.2
GenBank mRNA AI414273.1 -
AK041815.1 -
AK079617.1 -
AK158584.1 -
AK158782.1 -
AK164411.1 -
BC099872.1 -
BC099873.1 -
BC099874.1 -
BC099875.1 -
BC125237.1 -
BX512213.1 -
D38258.1 -
HM988990.1 -
S82023.1 -
U33535.1 -

View Full Product Details