Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001163530.1 | NP_001157002.1 |
NM_011164.2 | NP_035294.2 | |
GenBank mRNA | AK017520.1 | - |
AK017521.1 | - | |
AK017547.1 | - | |
AK017562.1 | - | |
AK017579.1 | - | |
AK017584.1 | - | |
AK077493.1 | - | |
AK133645.1 | - | |
AK133736.1 | - | |
AK133752.1 | - | |
AK161553.1 | - | |
AK161557.1 | - | |
AK161565.1 | - | |
AK161594.1 | - | |
AK161619.1 | - | |
BC061141.1 | - | |
X02892.1 | - | |
X04418.1 | - |