Gene Transcripts

Gene Symbol
Vegfc
Entrez Gene ID
22341
Gene Name
vascular endothelial growth factor C
Gene Aliases
AW228853, VEGF-C
Chromosome Location
Chr.8: 54077532 - 54186454 on Build GRCm38
Uni Gene
Mm.1402
Interrogated Sequence Translated Protein
RefSeq NM_009506.2 NP_033532.1
GenBank mRNA AK047844.1 -
AK216473.1 -
BC096377.1 -
EU862553.1 -
EU862554.1 -
EU862555.1 -
U58112.1 -
U73620.1 -

View Full Product Details