Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_008839.2 | NP_032865.2 |
GenBank mRNA | AK018164.1 | - |
AK029399.1 | - | |
AK039740.1 | - | |
AK051885.1 | - | |
AK053528.1 | - | |
AK078608.1 | - | |
AK151886.1 | - | |
AK164801.1 | - | |
BC089038.1 | - | |
BC130228.1 | - | |
U03279.1 | - |