Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001190830.1 | NP_001177759.1 |
NM_010589.6 | NP_034719.2 | |
GenBank mRNA | AK038268.1 | - |
AK088365.1 | - | |
AK142178.1 | - | |
AK156646.1 | - | |
AK164520.1 | - | |
AW456897.1 | - | |
BC027234.1 | - | |
BC105577.1 | - | |
BC131646.1 | - | |
BC131647.1 | - | |
L32955.1 | - | |
L33768.1 | - | |
L40172.1 | - |