Gene Transcripts

Gene Symbol
Ins
Entrez Gene ID
101719971
Gene Name
insulin
Gene Aliases
GW7_00389
Chromosome Location
Chr.Un|NW_004624767: 14999088 - 15000035 on Build HetGla_female_1.0
Interrogated Sequence Translated Protein
RefSeq XM_004852092.1 XP_004852149.1

View Full Product Details