Gene Transcripts

Gene Symbol
Copg2os2
Entrez Gene ID
100044236
Gene Name
coatomer protein complex, subunit gamma 2, opposite strand 2
Gene Aliases
AW049845, Copg2as2, Lb9, Mit1
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.22891
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002845.1 257
GenBank mRNA U20262.1 - 1615
U20264.1 - 257
U20265.1 - 1383

View Full Product Details