Gene Transcripts

Gene Symbol
9430060I03Rik
Entrez Gene ID
100037260
Gene Name
RIKEN cDNA 9430060I03 gene
Gene Aliases
Chromosome Location
Chr. 1: 92933526 - 92950590 on Build GRCm38
UniGene ID
Mm.442236
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015525.1 2 1013
GenBank mRNA AK137082.1 - 412
AK162459.1 - 412
AK170241.1 - 1 378
BC090260.1 - 2 1013

View Full Product Details