Gene Transcripts
- Gene Symbol
- 9430060I03Rik
- Entrez Gene ID
- 100037260
- Gene Name
- RIKEN cDNA 9430060I03 gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 92933526 - 92950590 on Build GRCm38
- UniGene ID
- Mm.442236
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015525.1 |
|
2 |
520 |
|
GenBank mRNA |
BC090260.1 |
- |
2 |
520 |
|
- Gene Symbol
- Capn10
- Entrez Gene ID
- 23830
- Gene Name
- calpain 10
- Gene Aliases
- AW049679, Capn8, mKIAA1845
- Chromosome Location
- Chr. 1: 92934408 - 92947948 on Build GRCm38
- UniGene ID
- Mm.294315
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BI986964.1 |
- |
1 |
313 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ922611.1 |
- |
|
521 |
|
Chr. - - [-] |
View Full Product Details