Gene Transcripts

Gene Symbol
Atad3aos
Entrez Gene ID
70448
Gene Name
ATPase family, AAA domain containing 3A, opposite strand
Gene Aliases
2610204G22Rik
Chromosome Location
Chr. 4: 155761192 - 155763560 on Build GRCm38
UniGene ID
Mm.70636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027971.1 2 145
NR_027972.1 2 145
GenBank mRNA AK011879.1 - 2 117
AK078249.1 - 2 146

View Full Product Details