Gene Transcripts

Gene Symbol
SPACA6
Entrez Gene ID
147650
Gene Name
sperm acrosome associated 6
Gene Aliases
LET7EH, LINC00085, NCRNA00085, SPACA6P
Chromosome Location
Chr. 19: 51688211 - 51712071 on Build GRCh38
UniGene ID
Hs.467174
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024330.1 1 501
GenBank mRNA AY358799.1 - 1 501
BC104208.2 - 1 480 40031428
BC104209.2 - 1 480

View Full Product Details