Gene Transcripts

Gene Symbol
ZNF503-AS2
Entrez Gene ID
100131213
Gene Name
ZNF503 antisense RNA 2
Gene Aliases
C10orf41, NCRNA00245
Chromosome Location
Chr. 10: 75401528 - 75408982 on Build GRCh38
UniGene ID
Hs.534598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024421.1 2 2062
NR_024422.1 2 1630
NR_110299.1 2 1626
NR_110300.1 2 1398
NR_110301.1 2 1515
NR_110302.1 2 1448

View Full Product Details