Gene Transcripts
- Gene Symbol
- WDFY3-AS2
- Entrez Gene ID
- 404201
- Gene Name
- WDFY3 antisense RNA 2
- Gene Aliases
- C4orf12, FBI4, NCRNA00247
- Chromosome Location
- Chr. 4: 84966818 - 85007015 on Build GRCh38
- UniGene ID
- Hs.720920
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015359.1 |
|
4 |
1302 |
|
GenBank mRNA |
AK124663.1 |
- |
1 |
303 |
|
AY250185.1 |
- |
7 |
1825 |
|
View Full Product Details