Gene Transcripts

Gene Symbol
TISP43
Entrez Gene ID
150527
Gene Name
uncharacterized LOC150527
Gene Aliases
Chromosome Location
Chr. 2: 130570846 - 130584161 on Build GRCh38
UniGene ID
Hs.125706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027313.1 3 1720
NR_027314.1 2 1544
GenBank mRNA BC064385.1 - 3 1720

Gene Symbol
LOC646743
Entrez Gene ID
646743
Gene Name
uncharacterized LOC646743
Gene Aliases
Chromosome Location
Chr. 2: 130536570 - 130549915 on Build GRCh38
UniGene ID
Hs.634887
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033930.1 3 1737

View Full Product Details