Gene Transcripts
- Gene Symbol
- TISP43
- Entrez Gene ID
- 150527
- Gene Name
- uncharacterized LOC150527
- Gene Aliases
- Chromosome Location
- Chr. 2: 130570846 - 130584161 on Build GRCh38
- UniGene ID
- Hs.125706
- Species
- Homo sapiens
- Gene Symbol
- LOC646743
- Entrez Gene ID
- 646743
- Gene Name
- uncharacterized LOC646743
- Gene Aliases
- Chromosome Location
- Chr. 2: 130536570 - 130549915 on Build GRCh38
- UniGene ID
- Hs.634887
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033930.1 |
|
3 |
1652 |
|
View Full Product Details