Gene Transcripts

Gene Symbol
LINC00176
Entrez Gene ID
284739
Gene Name
long intergenic non-protein coding RNA 176
Gene Aliases
NCRNA00176, PRR17
Chromosome Location
Chr. 20: 64034344 - 64039962 on Build GRCh38
UniGene ID
Hs.97840
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027686.1 2 3259
NR_027687.1 2 3259
GenBank mRNA AL157500.1 - 2 1940
BC033342.1 - 2 3259

View Full Product Details