Gene Transcripts
- Gene Symbol
- TMEM181
- Entrez Gene ID
- 57583
- Gene Name
- transmembrane protein 181
- Gene Aliases
- GPR178, KIAA1423
- Chromosome Location
- Chr. 6: 158536432 - 158635435 on Build GRCh38
- UniGene ID
- Hs.666596
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY927471.1 |
- |
LIT3489 AY927471 |
1 |
532 |
|
View Full Product Details