Gene Transcripts

Gene Symbol
TMEM181
Entrez Gene ID
57583
Gene Name
transmembrane protein 181
Gene Aliases
GPR178, KIAA1423
Chromosome Location
Chr. 6: 158536432 - 158635435 on Build GRCh38
UniGene ID
Hs.666596
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY927470.1 -
1 22
AY927471.1 - LIT3489
AY927471
1 22
AY927472.1 -
1 22

View Full Product Details