Gene Transcripts

Gene Symbol
LINC00837
Entrez Gene ID
100507605
Gene Name
long intergenic non-protein coding RNA 837
Gene Aliases
Chromosome Location
Chr. 10: 28789311 - 28796050 on Build GRCh38
UniGene ID
Hs.119207
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038374.1 3 2665
NR_038375.1 2 2563
NR_038376.1 2 2442
GenBank mRNA AL080132.1 - 1 1385
BC028422.1 - 2 2487

View Full Product Details