Gene Transcripts

Gene Symbol
LRCH3
Entrez Gene ID
84859
Gene Name
leucine rich repeats and calponin homology domain containing 3
Gene Aliases
Chromosome Location
Chr. 3: 197790855 - 197889346 on Build GRCh38
UniGene ID
Hs.518414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005269365.2 XP_005269422.1 20 8875
XM_005269367.2 XP_005269424.1 19 8803
XM_006713791.3 XP_006713854.1 21 5954
XM_006713792.3 XP_006713855.1 20 5882
XM_006713793.2 XP_006713856.1 20 5846
XM_017007352.1 XP_016862841.1 18 8749
GenBank mRNA AK125235.1 - 1 405


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA080686.1 - 1 262 Chr. 3 - 197886602 - 197887161 [+]
GenBankmRNA DA559512.1 - 1 405 Chr. 3 - 197886457 - 197887271 [+]
GenBankmRNA DA380104.1 - 1 344 Chr. 3 - 197886519 - 197887057 [+]

View Full Product Details