Gene Transcripts
- Gene Symbol
- RUFY2
- Entrez Gene ID
- 55680
- Gene Name
- RUN and FYVE domain containing 2
- Gene Aliases
- RABIP4R, ZFYVE13
- Chromosome Location
- Chr. 10: 68341107 - 68407302 on Build GRCh38
- UniGene ID
- Hs.653144
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC030097.1 |
- |
1 |
148 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB502513.1 |
- |
1 |
142 |
|
Chr. 10 - 68355213 - 68355687 [-] |
GenBankmRNA |
BG707400.1 |
- |
1 |
148 |
|
Chr. 10 - 68354953 - 68355693 [-] |
View Full Product Details