Gene Transcripts
- Gene Symbol
- FMOD
- Entrez Gene ID
- 2331
- Gene Name
- fibromodulin
- Gene Aliases
- FM, SLRR2E
- Chromosome Location
- Chr. 1: 203340621 - 203351429 on Build GRCh38
- UniGene ID
- Hs.519168
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098322.1 |
- |
1 |
699 |
|
View Full Product Details