Gene Transcripts

Gene Symbol
TMEM181
Entrez Gene ID
57583
Gene Name
transmembrane protein 181
Gene Aliases
GPR178, KIAA1423
Chromosome Location
Chr. 6: 158536432 - 158635435 on Build GRCh38
UniGene ID
Hs.666596
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011536003.2 XP_011534305.1 11 5257
GenBank mRNA AK023059.1 - 1 2228

View Full Product Details