Gene Transcripts
- Gene Symbol
- LOC100419583
- Entrez Gene ID
- 100419583
- Gene Name
- ring finger protein 4 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 15: 44728988 - 44732851 on Build GRCh38
- UniGene ID
- Hs.489254
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125376.1 |
|
2 |
1737 |
|
GenBank mRNA |
BC031266.1 |
- |
2 |
1603 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR592231.1 |
- |
1 |
277 |
|
Chr. 15 - 44732258 - 44733972 [+] |
GenBankmRNA |
BX378653.2 |
- |
1 |
277 |
|
Chr. 15 - 44732258 - 44733133 [+] |
View Full Product Details