Gene Transcripts

Gene Symbol
ECHDC2
Entrez Gene ID
55268
Gene Name
enoyl-CoA hydratase domain containing 2
Gene Aliases
Chromosome Location
Chr. 1: 52895910 - 52927212 on Build GRCh38
UniGene ID
Hs.476319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011541713.2 XP_011540015.1 1 3745
XM_011541720.2 XP_011540022.1 1 3745
XM_011541726.2 XP_011540028.1 1 3745
XM_011541727.2 XP_011540029.1 1 3745
XM_017001642.1 XP_016857131.1 1 3745
GenBank mRNA BX648522.1 - 1 748

View Full Product Details