Gene Transcripts

Gene Symbol
PRKG1-AS1
Entrez Gene ID
100506939
Gene Name
PRKG1 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 10: 52296848 - 52314128 on Build GRCh38
UniGene ID
Hs.584791
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038277.1 6 1314
GenBank mRNA BC050034.1 - 6 1316

Gene Symbol
PRKG1
Entrez Gene ID
5592
Gene Name
protein kinase, cGMP-dependent, type I
Gene Aliases
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha
Chromosome Location
Chr. 10: 50991151 - 52298350 on Build GRCh38
UniGene ID
Hs.407535
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BQ445850.1 - 1 580

View Full Product Details