Gene Transcripts

Gene Symbol
RALGPS1
Entrez Gene ID
9649
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases
RALGEF2, RALGPS1A
Chromosome Location
Chr. 9: 126914774 - 127223166 on Build GRCh38
UniGene ID
Hs.432842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017015348.1 XP_016870837.1 17 3559


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA436274.1 - 1 85 Chr. 9 - 127216504 - 127216991 [+]
GenBankmRNA CX873286.1 - 1 24 Chr. 9 - 127216565 - 127217322 [+]
GenBankmRNA AK023463.1 - 1 1352 Chr. 9 - 127215249 - 127217341 [+]

View Full Product Details