Gene Transcripts

Gene Symbol
CACTIN-AS1
Entrez Gene ID
404665
Gene Name
CACTIN antisense RNA 1
Gene Aliases
C19orf29-AS1, C19orf29OS
Chromosome Location
Chr. 19: 3607247 - 3613930 on Build GRCh38
UniGene ID
Hs.719726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK097999.1 - 3 437


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB116596.1 - 3 437 Chr. 19 - 3607247 - 3612077 [+]

View Full Product Details