Gene Transcripts
- Gene Symbol
- SSTR3
- Entrez Gene ID
- 6753
- Gene Name
- somatostatin receptor 3
- Gene Aliases
- SS-3-R, SS3-R, SS3R, SSR-28
- Chromosome Location
- Chr. 22: 37204237 - 37220451 on Build GRCh38
- UniGene ID
- Hs.225995
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BF514979.1 |
- |
1 |
404 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BC133662.1 |
- |
1 |
112 |
|
Chr. 22 - 37204521 - 37205449 [+] |
View Full Product Details