Gene Transcripts
- Gene Symbol
- Ccdc42os
- Entrez Gene ID
- 75151
- Gene Name
- coiled-coil domain containing 42, opposite strand
- Gene Aliases
- 4930535C22Rik
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.438576
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015975.1 |
- |
1 |
741 |
|
View Full Product Details