Gene Transcripts
- Gene Symbol
- 1700017G19Rik
- Entrez Gene ID
- 100040882
- Gene Name
- RIKEN cDNA 1700017G19 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 40504865 - 40522912 on Build GRCm38
- UniGene ID
- Mm.272869
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040445.1 |
|
4 |
1157 |
|
GenBank mRNA |
AK006064.1 |
- |
4 |
1158 |
|
View Full Product Details