Gene Transcripts
- Gene Symbol
- Il18rap
- Entrez Gene ID
- 16174
- Gene Name
- interleukin 18 receptor accessory protein
- Gene Aliases
- AcPL, IL-18R-beta, IL-18RAcP, IL-18Rbeta, IL-1RAcPL
- Chromosome Location
- Chr. 1: 40515362 - 40551707 on Build GRCm38
- UniGene ID
- Mm.20466
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK161196.1 |
- |
1 |
2087 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA096597.1 |
- |
1 |
213 |
|
Chr. 1 - 40550783 - 40551288 [+] |
View Full Product Details