Gene Transcripts
- Gene Symbol
- Hnrnph3
- Entrez Gene ID
- 432467
- Gene Name
- heterogeneous nuclear ribonucleoprotein H3
- Gene Aliases
- AA693301, AI666703, Hnrph3
- Chromosome Location
- Chr. 10: 63014650 - 63023889 on Build GRCm38
- UniGene ID
- Mm.274784
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC049652.1 |
- |
3 |
638 |
|
View Full Product Details