Gene Transcripts
- Gene Symbol
- 4930587E11Rik
- Entrez Gene ID
- 75893
- Gene Name
- RIKEN cDNA 4930587E11 gene
- Gene Aliases
- 4930523E13Rik
- Chromosome Location
- Chr. 6: 102265752 - 102337288 on Build GRCm38
- UniGene ID
- Mm.2968
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016361.1 |
- |
5 |
409 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY716007.1 |
- |
5 |
410 |
|
Chr. 6 - 102266121 - 102336677 [+] |
View Full Product Details