Gene Transcripts
- Gene Symbol
- Atp5l
- Entrez Gene ID
- 27425
- Gene Name
- ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G
- Gene Aliases
- 4933437C06Rik
- Chromosome Location
- Chr. 9: 44913248 - 44920742 on Build GRCm38
- UniGene ID
- Mm.14663
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017092.1 |
- |
|
631 |
|
View Full Product Details