Gene Transcripts
- Gene Symbol
- Ankra2
- Entrez Gene ID
- 68558
- Gene Name
- ankyrin repeat, family A (RFXANK-like), 2
- Gene Aliases
- 1110004M18Rik, Ankra
- Chromosome Location
- Chr. 13: 98263076 - 98274754 on Build GRCm38
- UniGene ID
- Mm.209642
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK171682.1 |
- |
1 |
1940 |
|
View Full Product Details