Gene Transcripts
- Gene Symbol
- Gm13990
- Entrez Gene ID
- 102634211
- Gene Name
- predicted gene 13990
- Gene Aliases
- OTTMUSG00000015246
- Chromosome Location
- Chr. 2: 116803444 - 116878668 on Build GRCm38
- UniGene ID
- Mm.406979
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054431.1 |
- |
4 |
646 |
|
View Full Product Details