Gene Transcripts
- Gene Symbol
- Mpzl2
- Entrez Gene ID
- 14012
- Gene Name
- myelin protein zero-like 2
- Gene Aliases
- Eva, Eva1
- Chromosome Location
- Chr. 9: 45040291 - 45054043 on Build GRCm38
- UniGene ID
- Mm.33240
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK044864.1 |
- |
|
311 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB635239.1 |
- |
1 |
491 |
|
Chr. 9 - 45054818 - 45055344 [-] |
GenBankmRNA |
AK040091.1 |
- |
|
491 |
|
Chr. - - [-] |
GenBankmRNA |
BB204483.2 |
- |
1 |
505 |
|
Chr. 9 - 45054823 - 45055344 [-] |
View Full Product Details