Gene Transcripts
- Gene Symbol
- Plxna4os2
- Entrez Gene ID
- 330283
- Gene Name
- plexin A4, opposite strand 2
- Gene Aliases
- 9430029A11Rik
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.242174
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034724.1 |
- |
3 |
1067 |
|
View Full Product Details