Gene Transcripts
- Gene Symbol
- 2900084C01Rik
- Entrez Gene ID
- 73081
- Gene Name
- RIKEN cDNA 2900084C01 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013816.1 |
- |
1 |
195 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY712781.1 |
- |
1 |
187 |
|
Chr. 10 - 13965778 - 13965979 [+] |
View Full Product Details