Gene Transcripts
- Gene Symbol
- Rela
- Entrez Gene ID
- 19697
- Gene Name
- v-rel reticuloendotheliosis viral oncogene homolog A (avian)
- Gene Aliases
- p65
- Chromosome Location
- Chr. 19: 5637440 - 5648130 on Build GRCm38
- UniGene ID
- Mm.249966
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK154938.1 |
- |
|
1337 |
|
View Full Product Details