Gene Transcripts
- Gene Symbol
- 8430416G17Rik
- Entrez Gene ID
- 71469
- Gene Name
- RIKEN cDNA 8430416G17 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.100172
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC038171.1 |
- |
1 |
18 |
|
- Gene Symbol
- Gm4013
- Entrez Gene ID
- 100042757
- Gene Name
- predicted gene 4013
- Gene Aliases
- Chromosome Location
- Chr. 18: 42274380 - 42275197 on Build GRCm38
- UniGene ID
- Mm.100172
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033452.1 |
|
1 |
28 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB318895.1 |
- |
1 |
15 |
|
Chr. 18 - 42274396 - 42275181 [-] |
GenBankmRNA |
BE448321.1 |
- |
1 |
18 |
|
Chr. 18 - 42274391 - 42275187 [-] |
GenBankmRNA |
BY099906.1 |
- |
1 |
28 |
|
Chr. 18 - 42274441 - 42275197 [-] |
GenBankmRNA |
AW990362.1 |
- |
1 |
12 |
|
Chr. 18 - 42265000 - 42275181 [-] |
View Full Product Details