Gene Transcripts
- Gene Symbol
- 2310069B03Rik
- Entrez Gene ID
- 69652
- Gene Name
- RIKEN cDNA 2310069B03 gene
- Gene Aliases
- Chromosome Location
- Chr. 6: 82877866 - 82881853 on Build GRCm38
- UniGene ID
- Mm.96357
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040520.1 |
|
4 |
1183 |
|
GenBank mRNA |
AK078808.1 |
- |
4 |
1184 |
|
View Full Product Details