Gene Transcripts
- Gene Symbol
- Atp6v0a2
- Entrez Gene ID
- 21871
- Gene Name
- ATPase, H+ transporting, lysosomal V0 subunit A2
- Gene Aliases
- 8430408C20Rik, AI385560, ATP6a2, AW489264, Atp6n1d, Atp6n2, C76904, ISF, J6B7, SHIF, Stv1, TJ6M, TJ6s, Tj6, V-ATPase 116 kDa, V-ATPase a2
- Chromosome Location
- Chr. 5: 124629052 - 124646816 on Build GRCm38
- UniGene ID
- Mm.1158
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK160318.1 |
- |
1 |
650 |
|
BC112905.1 |
- |
|
5009 |
3670722 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CO045780.1 |
- |
1 |
713 |
|
Chr. 5 - 124723448 - 124724271 [+] |
GenBankmRNA |
BY643209.1 |
- |
1 |
137 |
|
Chr. 5 - 124724025 - 124724449 [+] |
GenBankmRNA |
BX528274.1 |
- |
1 |
242 |
|
Chr. 5 - 124723920 - 124724450 [+] |
GenBankmRNA |
AA419856.1 |
- |
1 |
282 |
|
Chr. 5 - 124723877 - 124724405 [+] |
GenBankmRNA |
CK634097.1 |
- |
1 |
200 |
|
Chr. 5 - 124723961 - 124724450 [+] |
View Full Product Details